siRNA Details
| Gene Symbol: Nfib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113209.2 | NP_001106680.1 | 5 | 1770 | ||
| NM_001113210.2 | NP_001106681.1 | 5 | 1770 | |||
| NM_001286127.1 | NP_001273056.1 | 5 | 1770 | |||
| NM_001286131.1 | NP_001273060.1 | 5 | 1770 | |||
| NM_008687.6 | NP_032713.3 | 5 | 1770 | |||
| GenBank mRNA | AK034261.1 | - | 5 | 1302 | ||
| AK083167.1 | - | 5 | 1115 | |||
| AK084941.1 | - | 5 | 1375 | |||
| AK087480.1 | - | 5 | 1302 | |||
| AK145267.1 | - | 4 | 476 | |||
| AK146480.1 | - | 5 | 1287 | |||
| AK148573.1 | - | 5 | 1758 | |||
| AY035852.1 | - | 5 | 775 | |||
| AY035853.1 | - | 5 | 775 | |||
| BC014290.1 | - | 5 | 807 | 4038233 | ||
| BC023366.1 | - | 5 | 907 | |||
| BC062908.1 | - | 5 | 1092 | 6815775 | ||
| BE862566.1 | - | 4 | 421 | |||
| U57634.1 | - | 5 | 976 | |||
| Y07685.1 | - | 5 | 1702 | |||
| Y07686.1 | - | 5 | 1282 | |||
| Y07687.1 | - | 5 | 1763 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031566, XM_008763762, XM_006238335, XM_006238336, XM_006238339, and others.

Nfib
Genomic Map