siRNA Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008689.2 | NP_032715.2 | 7 | 782 | ||
| GenBank mRNA | AB119195.1 | - | 5 | 280 | ||
| AB221614.1 | - | 5 | 287 | |||
| AK052726.1 | - | 2 | 90 | |||
| AY423849.1 | - | 2 | 121 | |||
| AY521462.1 | - | 5 | 280 | |||
| AY521463.1 | - | 5 | 280 | |||
| BC050841.1 | - | 7 | 782 | |||
| BC138535.1 | - | 6 | 359 | 8861555 | ||
| BC138536.1 | - | 6 | 359 | 8861556 | ||
| BC145317.1 | - | 7 | 376 | |||
| M57999.1 | - | 7 | 571 | |||

Nfkb1
Genomic Map