siRNA Details
| Gene Symbol: Nefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008691.2 | NP_032717.2 | 2 | 1324 | ||
| GenBank mRNA | AK018234.1 | - | 2 | 1163 | ||
| AK051296.1 | - | 2 | 1167 | |||
| AK051696.1 | - | 2 | 1164 | |||
| AK080794.1 | - | 2 | 1168 | |||
| AK164318.1 | - | 2 | 1167 | |||
| AK165041.1 | - | 2 | 1167 | |||
| BC052352.1 | - | 2 | 1166 | |||
| BC058333.1 | - | 2 | 1157 | |||
| BC119602.2 | - | 2 | 1132 | |||
| BC128564.1 | - | 2 | 1127 | |||
| DQ201636.1 | - | 2 | 1142 | |||
| M20481.1 | - | 2 | 170 | |||

Nefm
Genomic Map