siRNA Details
| Gene Symbol: Nfyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048168.2 | NP_001041633.1 | 4 | 373 | ||
| NM_008692.4 | NP_032718.2 | 4 | 446 | |||
| GenBank mRNA | AB006181.1 | - | 3 | 226 | ||
| AK011327.1 | - | 4 | 403 | |||
| BC020117.1 | - | 4 | 372 | 4009737 | ||
| BC053723.1 | - | 4 | 376 | 30053020 | ||
| BC085261.1 | - | 4 | 372 | 6397760 | ||
| BM461330.1 | - | 2 | 77 | |||
| CF172077.1 | - | 4 | 369 | |||
| CJ092564.1 | - | 4 | 423 | |||
| DV650959.1 | - | 4 | 385 | |||
| U62297.1 | - | 3 | 226 | |||

Nfyc
Genomic Map