siRNA Details
| Gene Symbol: Klk1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008693.2 | NP_032719.1 | 3 | 328 | ||
| GenBank mRNA | BC132657.1 | - | 3 | 328 | 40130934 | |
| BC132659.1 | - | 3 | 328 | 40130936 | ||
| X01389.1 | - | 3 | 289 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk1b3| Gene Symbol: Klk1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008693.2 | NP_032719.1 | 3 | 328 | ||
| GenBank mRNA | BC132657.1 | - | 3 | 328 | 40130934 | |
| BC132659.1 | - | 3 | 328 | 40130936 | ||
| X01389.1 | - | 3 | 289 | |||