siRNA Details
| Gene Symbol: Nid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010917.2 | NP_035047.2 | 3 | 804 | ||
| GenBank mRNA | AK041633.1 | - | 3 | 740 | ||
| AK049585.1 | - | 3 | 740 | |||
| AK084876.1 | - | 3 | 743 | |||
| AK144878.1 | - | 3 | 769 | |||
| AK166779.1 | - | 3 | 740 | |||
| BC052939.1 | - | 3 | 799 | |||
| BC131669.1 | - | 3 | 715 | 40110859 | ||
| X14194.1 | - | 3 | 715 | |||
| X14480.1 | - | 3 | 725 | |||

Nid1
Genomic Map