siRNA Details
| Gene Symbol: Nktr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010918.2 | NP_035048.3 | 9, 10 | 910 | ||
| GenBank mRNA | AK035582.1 | - | 8, 9 | 736 | ||
| AK144214.1 | - | 9, 10 | 821 | |||
| BC048414.1 | - | 10, 11 | 900 | |||
| L04289.2 | - | 9, 10 | 915 | |||

Nktr
Genomic Map