siRNA Details
Gene Symbol: Nktr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010918.2 | NP_035048.3 | 8 | 597 | ||
GenBank mRNA | AK035582.1 | - | 7 | 498 | ||
AK144214.1 | - | 8 | 508 | |||
AK163916.1 | - | 10 | 2327 | |||
BC048414.1 | - | 9 | 587 | |||
L04289.2 | - | 8 | 602 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
