siRNA Details
Gene Symbol: Nkx6-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144955.2 | NP_659204.1 | 2 | 1164 | ||
GenBank mRNA | AF357883.1 | - | 2 | 1164 | ||
BC138019.1 | - | 2 | 1070 | 8861038 | ||
BC138020.1 | - | 2 | 1070 | 8861039 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031737, AF004431.
Panther Classification:
Gene Ontology Categories:
