siRNA Details
| Gene Symbol: Nqo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163239.1 | NP_001156711.1 | 4 | 471 | ||
| NM_001163241.1 | NP_001156713.1 | 5 | 559 | |||
| NM_001163242.1 | NP_001156714.1 | 5 | 488 | |||
| NM_020282.3 | NP_064678.1 | 6 | 641 | |||
| GenBank mRNA | AF252260.1 | - | 5 | 487 | ||
| AK008138.1 | - | 4 | 443 | |||
| AK008465.1 | - | 4 | 446 | |||
| AK010842.1 | - | 5 | 531 | |||
| AK049781.1 | - | 5 | 497 | |||
| BC027629.1 | - | 6 | 549 | 1225122 | ||

Nqo2
Genomic Map