siRNA Details
| Gene Symbol: Nmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290368.1 | NP_001277297.1 | 1 | 161 | ||
| NM_001290369.1 | NP_001277298.1 | 1 | 161 | |||
| NM_001290370.1 | NP_001277299.1 | 1 | 161 | |||
| NM_008708.2 | NP_032734.1 | 1 | 161 | |||
| GenBank mRNA | AF043327.1 | - | 1 | 85 | ||
| AK044213.1 | - | 1 | 93 | |||
| AK134521.1 | - | 1 | 125 | |||
| AK161490.1 | - | 1 | 161 | |||
| AK168171.1 | - | 1 | 129 | |||
| AK168190.1 | - | 1 | 129 | |||
| BC037647.1 | - | 1 | 108 | 5253732 | ||

Nmt2
Genomic Map