siRNA Details
| Gene Symbol: Nnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291128.1 | NP_001278057.1 | 3 | 688 | ||
| NM_001291129.1 | NP_001278058.1 | 3 | 676 | |||
| NM_001291130.1 | NP_001278059.1 | 3 | 627 | |||
| NM_010923.3 | NP_035053.1 | 3 | 681 | |||
| NM_180960.3 | NP_851291.1 | 2 | 600 | |||
| GenBank mRNA | AB004048.1 | - | 3 | 887 | ||
| AK003004.1 | - | 3 | 645 | |||
| AK160626.1 | - | 3 | 649 | |||
| AU079843.1 | - | 2 | 567 | |||
| AY644389.1 | - | 1 | 410 | |||
| BC036984.1 | - | 3 | 639 | 4981517 | ||
| DV062509.1 | - | 3 | 699 | |||

Nnat
Genomic Map