siRNA Details
| Gene Symbol: Nnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010924.2 | NP_035054.1 | 1 | 224 | ||
| GenBank mRNA | CF732455.1 | - | 1 | 224 | ||
| U86105.1 | - | 1 | 271 | |||
| U86107.1 | - | 1 | 271 | |||
| U89377.1 | - | 1 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nnmt| Gene Symbol: Nnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010924.2 | NP_035054.1 | 1 | 224 | ||
| GenBank mRNA | CF732455.1 | - | 1 | 224 | ||
| U86105.1 | - | 1 | 271 | |||
| U86107.1 | - | 1 | 271 | |||
| U89377.1 | - | 1 | 271 | |||