siRNA Details
| Gene Symbol: Rrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010925.2 | NP_035055.2 | 2 | 247 | ||
| GenBank mRNA | AK077540.1 | - | 2 | 172 | ||
| BC065993.1 | - | 2 | 209 | 5684672 | ||
| BC065994.1 | - | 2 | 167 | 5685269 | ||
| BY106122.1 | - | 2 | 247 | |||
| U79773.1 | - | 2 | 199 | |||
| U79774.1 | - | 2 | 199 | |||

Rrp1
Genomic Map