siRNA Details
| Gene Symbol: Nnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008710.3 | NP_032736.2 | 1224 | |||
| GenBank mRNA | AK050478.1 | - | 1129 | |||
| AK088044.1 | - | 1206 | ||||
| AK090115.1 | - | 1227 | ||||
| AK129006.1 | - | 1201 | ||||
| AK159694.1 | - | 1210 | ||||
| AK168736.1 | - | 1007 | ||||
| AK198291.1 | - | 353 | ||||
| BC008518.1 | - | 1187 | 3601702 | |||
| Z49204.1 | - | 1131 | ||||

Nnt
Genomic Map