siRNA Details
| Gene Symbol: Emc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010926.5 | NP_035056.1 | 4, 5 | 604 | ||
| GenBank mRNA | AF052621.1 | - | 4, 5 | 606 | ||
| AK042704.1 | - | 4, 5 | 472 | |||
| AK076710.1 | - | 6, 7 | 1473 | |||
| AK143131.1 | - | 4, 5 | 565 | |||
| AK148190.1 | - | 4, 5 | 1115 | |||
| AK161438.1 | - | 4, 5 | 580 | |||
| AK164130.1 | - | 4, 5 | 605 | |||
| BC009103.1 | - | 4, 5 | 560 | 3482942 | ||

Emc8
Genomic Map