siRNA Details
| Gene Symbol: Emc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010926.5 | NP_035056.1 | 5 | 678 | ||
| GenBank mRNA | AF052621.1 | - | 5 | 680 | ||
| AK042704.1 | - | 5 | 546 | |||
| AK076710.1 | - | 7 | 1547 | |||
| AK143131.1 | - | 5 | 639 | |||
| AK148190.1 | - | 5 | 1189 | |||
| AK161438.1 | - | 5 | 654 | |||
| AK164130.1 | - | 5 | 679 | |||
| BC009103.1 | - | 5 | 634 | 3482942 | ||

Emc8
Genomic Map