siRNA Details
| Gene Symbol: Nos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC066101.1 | - | 23 | 3993 | ||
| BC083183.1 | - | 23 | 3993 | |||
| D14552.1 | - | 22 | 3620 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nos1| Gene Symbol: Nos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC066101.1 | - | 23 | 3993 | ||
| BC083183.1 | - | 23 | 3993 | |||
| D14552.1 | - | 22 | 3620 | |||