siRNA Details
| Gene Symbol: Ints6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008715.2 | NP_032741.2 | 4 | 853 | ||
| GenBank mRNA | AK149711.1 | - | 4 | 881 | ||
| AK172178.1 | - | 4 | 881 | |||
| AK186108.1 | - | 2 | 185 | |||
| BC059263.1 | - | 4 | 853 | 6409580 | ||

Ints6
Genomic Map