siRNA Details
| Gene Symbol: Npc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008720.2 | NP_032746.2 | 8 | 1278 | ||
| GenBank mRNA | AF003348.1 | - | 8 | 1255 | ||
| AK154282.1 | - | 8 | 1198 | |||
| AK155379.1 | - | 8 | 1198 | |||
| BC052437.1 | - | 8 | 1278 | 6405214 | ||
| BC054539.1 | - | 8 | 1278 | 6405214 | ||

Npc1
Genomic Map