siRNA Details
| Gene Symbol: Npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039181.1 | NP_001034270.1 | 6 | 1416 | ||
| NM_001286395.1 | NP_001273324.1 | 7 | 1032 | |||
| NM_008728.2 | NP_032754.2 | 6 | 1416 | |||
| GenBank mRNA | AA763613.1 | - | 1 | 96 | ||
| AK079196.1 | - | 7 | 949 | |||
| BC055897.1 | - | 6 | 1416 | 4019152 | ||
| BQ831432.1 | - | 3 | 267 | |||
| D78175.1 | - | 6 | 1381 | |||

Npr3
Genomic Map