siRNA Details
| Gene Symbol: Npy1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010934.4 | NP_035064.1 | 2 | 927 | ||
| GenBank mRNA | AK132397.1 | - | 2 | 928 | ||
| D63818.1 | - | 1 | 762 | |||
| D63819.1 | - | 884 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC051420.1 | - | 820 | 6490858 | Chr. - - [-] on Build | |
More Information
Set Membership: |
|

Npy1r
Genomic Map