siRNA Details
| Gene Symbol: Slc11a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013612.2 | NP_038640.2 | 7 | 708 | ||
| GenBank mRNA | AK090303.1 | - | 7 | 709 | ||
| AK144237.1 | - | 7 | 691 | |||
| AK171343.1 | - | 6 | 944 | |||
| AK171393.1 | - | 7 | 689 | |||
| BC109137.1 | - | 7 | 622 | 40039390 | ||
| BC109138.1 | - | 7 | 622 | 40039397 | ||
| L13732.1 | - | 7 | 686 | |||
| X75355.1 | - | 7 | 667 | |||

Slc11a1
Genomic Map