siRNA Details
| Gene Symbol: Nrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164226.1 | NP_001157698.1 | 8 | 1083 | ||
| NM_001164227.1 | NP_001157699.1 | 8 | 1083 | |||
| NM_001164228.1 | NP_001157700.1 | 8 | 1080 | |||
| NM_001164229.1 | NP_001157701.1 | 9 | 1157 | |||
| NM_001164230.1 | NP_001157702.1 | 8 | 1080 | |||
| NM_010938.4 | NP_035068.3 | 8 | 1080 | |||
| GenBank mRNA | AF098077.1 | - | 8 | 1015 | ||
| AK014494.1 | - | 8 | 1044 | |||
| AK029034.1 | - | 8 | 1061 | |||
| AK031103.1 | - | 3 | 284 | |||
| AK037697.1 | - | 8 | 1041 | |||
| AK050861.1 | - | 8 | 997 | |||
| AK082580.1 | - | 8 | 1081 | |||
| AK134483.1 | - | 8 | 997 | |||
| AK135673.1 | - | 9 | 1160 | |||
| BC005410.1 | - | 7 | 901 | 3496661 | ||

Nrf1
Genomic Map