siRNA Details
| Gene Symbol: Nrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008737.2 | NP_032763.2 | 9 | 2033 | ||
| GenBank mRNA | AK082992.1 | - | 9 | 2086 | ||
| AK141964.1 | - | 9 | 1825 | |||
| AK148372.1 | - | 9 | 2009 | |||
| AK149752.1 | - | 4 | 614 | |||
| AK159219.1 | - | 9 | 1824 | |||
| AK179138.1 | - | 1 | 140 | |||
| BC060129.1 | - | 9 | 1813 | 6409596 | ||
| D50086.1 | - | 9 | 1893 | |||

Nrp1
Genomic Map