siRNA Details
| Gene Symbol: Nsdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010941.3 | NP_035071.3 | 6 | 845 | ||
| GenBank mRNA | AF100198.1 | - | 6 | 845 | ||
| AK013945.1 | - | 6 | 728 | |||
| AK088026.1 | - | 6 | 726 | |||
| AK140930.1 | - | 6 | 726 | |||
| BC019945.1 | - | 6 | 714 | 3989811 | ||
| BC052834.1 | - | 6 | 806 | 30048305 | ||
| KF356400.1 | - | 5 | 630 | |||
| KF356401.1 | - | 5 | 630 | |||

Nsdhl
Genomic Map