siRNA Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008740.4 | NP_032766.2 | 11 | 1207 | ||
| GenBank mRNA | AK049281.1 | - | 11 | 1177 | ||
| AK085086.1 | - | 11 | 1161 | |||
| AK153905.1 | - | 11 | 1176 | |||
| AK216127.1 | - | 2 | 110 | |||
| BC006627.1 | - | 11 | 1212 | 3968733 | ||
| BC019167.1 | - | 11 | 1160 | 4506351 | ||
| U10120.1 | - | 11 | 1165 | |||

Nsf
Genomic Map