siRNA Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008740.4 | NP_032766.2 | 7, 8 | 658 | ||
| GenBank mRNA | AK042341.1 | - | 8, 9 | 676 | ||
| AK049281.1 | - | 7, 8 | 628 | |||
| AK051430.1 | - | 7, 8 | 627 | |||
| AK085086.1 | - | 7, 8 | 612 | |||
| AK141986.1 | - | 7, 8 | 628 | |||
| AK153905.1 | - | 7, 8 | 627 | |||
| AK208406.1 | - | 2, 3 | 98 | |||
| BC006627.1 | - | 7, 8 | 663 | 3968733 | ||
| BC019167.1 | - | 7, 8 | 611 | 4506351 | ||
| U10120.1 | - | 7, 8 | 616 | |||

Nsf
Genomic Map