siRNA Details
| Gene Symbol: Numb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136075.2 | NP_001129547.1 | 10 | 1716 | ||
| NM_001272055.1 | NP_001258984.1 | 9 | 1683 | |||
| NM_001272056.1 | NP_001258985.1 | 10 | 1766 | |||
| NM_010949.2 | NP_035079.1 | 9 | 1569 | |||
| GenBank mRNA | AF169191.1 | - | 9 | 1668 | ||
| AF169192.1 | - | 10 | 1701 | |||
| AF170709.1 | - | 9 | 1554 | |||
| AK004553.1 | - | 11 | 1791 | |||
| BC033459.1 | - | 11 | 1807 | 3991630 | ||
| BC050108.1 | - | 13 | 1839 | |||
| U70674.1 | - | 10 | 1687 | |||

Numb
Genomic Map