siRNA Details
| Gene Symbol: Nr4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139509.1 | NP_001132981.1 | 3 | 445 | ||
| NM_013613.2 | NP_038641.1 | 3 | 591 | |||
| GenBank mRNA | AK087495.1 | - | 4 | 466 | ||
| AK158607.1 | - | 3 | 592 | |||
| BC137715.1 | - | 3 | 903 | 8860734 | ||
| BC144854.1 | - | 3 | 1119 | |||
| S53744.1 | - | 3 | 482 | |||

Nr4a2
Genomic Map