siRNA Details
| Gene Symbol: Nxph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008752.2 | NP_032778.1 | 2 | 644 | ||
| GenBank mRNA | AK038402.1 | - | 2 | 645 | ||
| BC106863.1 | - | 2 | 614 | 40047329 | ||
| BC106864.2 | - | 2 | 614 | 40047331 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nxph2| Gene Symbol: Nxph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008752.2 | NP_032778.1 | 2 | 644 | ||
| GenBank mRNA | AK038402.1 | - | 2 | 645 | ||
| BC106863.1 | - | 2 | 614 | 40047329 | ||
| BC106864.2 | - | 2 | 614 | 40047331 | ||