siRNA Details
| Gene Symbol: ENDOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004435.2 | NP_004426.2 | 2 | 721 | ||
| XM_011518347.2 | XP_011516649.1 | 2 | 819 | |||
| GenBank mRNA | AK130973.1 | - | 2 | 202 | ||
| BC004922.2 | - | 2 | 596 | 3604703 | ||
| BC016351.1 | - | 2 | 751 | 4080883 | ||
| X79444.1 | - | 2 | 702 | |||

ENDOG
Genomic Map