siRNA Details
| Gene Symbol: Obp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008754.2 | NP_032780.2 | 5 | 446 | ||
| GenBank mRNA | AK017455.1 | - | 5 | 447 | ||
| BC125514.1 | - | 5 | 433 | 40130029 | ||
| BC125516.1 | - | 5 | 433 | 40130031 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Obp1a| Gene Symbol: Obp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008754.2 | NP_032780.2 | 5 | 446 | ||
| GenBank mRNA | AK017455.1 | - | 5 | 447 | ||
| BC125514.1 | - | 5 | 433 | 40130029 | ||
| BC125516.1 | - | 5 | 433 | 40130031 | ||