siRNA Details
| Gene Symbol: Ocln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008756.2 | NP_032782.1 | 3 | 690 | ||
| GenBank mRNA | AK019880.1 | - | 3 | 642 | ||
| AK033532.1 | - | 3 | 623 | |||
| AK042576.1 | - | 3 | 690 | |||
| BC138679.1 | - | 3 | 673 | 8861700 | ||
| BC138680.1 | - | 3 | 673 | 8861701 | ||
| BC145371.1 | - | 3 | 688 | 9053929 | ||
| U49185.1 | - | 3 | 733 | |||

Ocln
Genomic Map