siRNA Details
| Gene Symbol: Ogn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008760.4 | NP_032786.1 | 7 | 868 | ||
| GenBank mRNA | AK028300.1 | - | 7 | 895 | ||
| AK053650.1 | - | 7 | 869 | |||
| AK160760.1 | - | 7 | 890 | |||
| AK163772.1 | - | 6 | 820 | |||
| BC021939.1 | - | 6 | 771 | 5067073 | ||
| D31951.1 | - | 6 | 781 | |||

Ogn
Genomic Map