siRNA Details
| Gene Symbol: Olfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010983.2 | NP_035113.1 | 2 | 388 | ||
| GenBank mRNA | BC128014.1 | - | 1 | 126 | 40136059 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr2| Gene Symbol: Olfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010983.2 | NP_035113.1 | 2 | 388 | ||
| GenBank mRNA | BC128014.1 | - | 1 | 126 | 40136059 | |