siRNA Details
| Gene Symbol: Olfr44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146830.2 | NP_667041.2 | 2 | 919 | ||
| GenBank mRNA | CB173511.1 | - | 1 | 426 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr44| Gene Symbol: Olfr44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146830.2 | NP_667041.2 | 2 | 919 | ||
| GenBank mRNA | CB173511.1 | - | 1 | 426 | ||