siRNA Details
| Gene Symbol: Olfr45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146963.1 | NP_667174.1 | 1 | 75 | ||
| GenBank mRNA | BC125298.1 | - | 1 | 98 | 40129813 | |
| BC125300.1 | - | 1 | 98 | 40129815 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr45| Gene Symbol: Olfr45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146963.1 | NP_667174.1 | 1 | 75 | ||
| GenBank mRNA | BC125298.1 | - | 1 | 98 | 40129813 | |
| BC125300.1 | - | 1 | 98 | 40129815 | ||