siRNA Details
Gene Symbol: Olfr51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146909.1 | NP_667120.1 | 1 | 53 | ||
GenBank mRNA | BC111814.1 | - | 1 | 52 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146909.1 | NP_667120.1 | 1 | 53 | ||
GenBank mRNA | BC111814.1 | - | 1 | 52 |