siRNA Details
| Gene Symbol: Olfr56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010999.2 | NP_035129.1 | 6 | 1203 | ||
| GenBank mRNA | AF102533.1 | - | 1 | 275 | ||
| AF102535.1 | - | 1 | 275 | |||
| AF102537.1 | - | 1 | 275 | |||
| AK053352.1 | - | 6 | 1203 | |||
| BC141219.1 | - | 5 | 1034 | 9055831 | ||
| BC147437.1 | - | 5 | 1148 | 9056030 | ||

Olfr56
Genomic Map