siRNA Details
Gene Symbol: Olfr1351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147040.1 | NP_667251.1 | 1 | 457 | ||
GenBank mRNA | AF364956.1 | - | 1 | 112 | ||
BC104112.1 | - | 1 | 457 | 40043939 | ||
BC104113.1 | - | 1 | 456 |
Gene Symbol: Olfr57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147041.2 | NP_667252.1 | 1 | 532 | ||
GenBank mRNA | AF042359.1 | - | 1 | 112 | ||
AF102534.1 | - | 1 | 265 | |||
BC141307.1 | - | 1 | 536 | 9055919 |
