siRNA Details
| Gene Symbol: Omt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111286.2 | NP_001104756.1 | 2 | 742 | ||
| NM_001164523.1 | NP_001157995.1 | 2 | 121 | |||
| GenBank mRNA | AA619817.1 | - | 2 | 110 | ||
| AA693017.1 | - | 2 | 121 | |||
| AK135846.1 | - | 2 | 168 | |||
| AK139546.1 | - | 2 | 710 | |||
| BB698495.1 | - | 2 | 52 | |||
| DV647767.1 | - | 2 | 174 | |||
| DV648064.1 | - | 2 | 186 | |||
| S81934.1 | - | 3 | 278 | |||

Omt2a
Genomic Map