siRNA Details
| Gene Symbol: Orm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013623.2 | NP_038651.2 | 5 | 514 | ||
| GenBank mRNA | BC141235.1 | - | 5 | 518 | 9055847 | |
| BC145559.1 | - | 5 | 518 | 9054123 | ||
| BC145560.1 | - | 5 | 548 | |||
| BC171932.1 | - | 4 | 375 | |||

Orm3
Genomic Map