siRNA Details
| Gene Symbol: Otx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286481.1 | NP_001273410.1 | 5 | 745 | ||
| NM_001286482.1 | NP_001273411.1 | 5 | 721 | |||
| NM_001286483.1 | NP_001273412.1 | 4 | 561 | |||
| NM_144841.4 | NP_659090.1 | 3 | 643 | |||
| GenBank mRNA | AK053741.1 | - | 4 | 558 | ||
| AK081535.1 | - | 4 | 562 | |||
| AK087527.1 | - | 4 | 559 | |||
| BC017609.1 | - | 3 | 643 | 4527414 | ||
| BC027104.1 | - | 5 | 603 | 5359966 | ||
| BC029667.1 | - | 5 | 607 | 5362013 | ||
| BQ959742.1 | - | 5 | 721 | |||
| CO424557.1 | - | 5 | 646 | |||

Otx2
Genomic Map