siRNA Details
| Gene Symbol: P2rx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011026.2 | NP_035156.2 | 8 | 857 | ||
| GenBank mRNA | AF089751.1 | - | 8 | 791 | ||
| AF089752.1 | - | 7 | 710 | |||
| AJ251459.1 | - | 8 | 771 | |||
| AJ251460.1 | - | 7 | 690 | |||
| AJ251461.1 | - | 8 | 771 | |||
| AJ251462.1 | - | 7 | 690 | |||
| AK150351.1 | - | 8 | 847 | |||
| AK162820.1 | - | 8 | 823 | |||
| AK163104.1 | - | 8 | 845 | |||
| BC002099.1 | - | 8 | 831 | |||
| BC005597.1 | - | 7 | 724 | 3482458 | ||
| JX417451.1 | - | 8 | 817 | |||
| JX674050.1 | - | 4 | 425 | |||

P2rx4
Genomic Map