siRNA Details
| Gene Symbol: P2ry1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282016.1 | NP_001268945.1 | 1 | 1732 | ||
| NM_008772.5 | NP_032798.1 | 1 | 1732 | |||
| GenBank mRNA | AK030759.1 | - | 1 | 1721 | ||
| AK033660.1 | - | 1 | 1718 | |||
| AK036611.1 | - | 1 | 1723 | |||
| BC138634.1 | - | 1 | 1593 | 8861654 | ||
| BC138635.1 | - | 1 | 1593 | 8861655 | ||
| EF584663.1 | - | 1 | 84 | |||
| U22829.1 | - | 1 | 1106 | |||
More Information
Set Membership: |
|

P2ry1
Genomic Map