siRNA Details
| Gene Symbol: P4ha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136076.2 | NP_001129548.1 | 8 | 1094 | ||
| NM_011031.2 | NP_035161.2 | 8 | 1094 | |||
| GenBank mRNA | AK053504.1 | - | 8 | 1054 | ||
| AK144124.1 | - | 8 | 1041 | |||
| AK145535.1 | - | 8 | 1021 | |||
| AK195504.1 | - | 1 | 22 | |||
| BC018411.1 | - | 8 | 1006 | 3157481 | ||
| U16163.1 | - | 7 | 981 | |||

P4ha2
Genomic Map