siRNA Details
| Gene Symbol: P4ha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136076.2 | NP_001129548.1 | 11, 12 | 1518 | ||
| NM_011031.2 | NP_035161.2 | 11, 12 | 1518 | |||
| GenBank mRNA | AK053504.1 | - | 11, 12 | 1478 | ||
| AK142689.1 | - | 3, 4 | 213 | |||
| AK144124.1 | - | 11, 12 | 1465 | |||
| AK145535.1 | - | 11, 12 | 1445 | |||
| BC018411.1 | - | 11, 12 | 1430 | 3157481 | ||
| CD563665.1 | - | 4, 5 | 376 | |||
| U16163.1 | - | 10, 11 | 1405 | |||

P4ha2
Genomic Map