siRNA Details
| Gene Symbol: Bloc1s6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019788.3 | NP_062762.1 | 1, 2 | 207 | ||
| GenBank mRNA | AK038381.1 | - | 1, 2 | 185 | ||
| AK077720.1 | - | 1, 2 | 189 | |||
| AK147116.1 | - | 1, 2 | 210 | |||
| AK160580.1 | - | 1, 2 | 177 | |||
| BY015259.1 | - | 1, 2 | 208 | |||

Bloc1s6
Genomic Map