siRNA Details
| Gene Symbol: Pabpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011033.2 | NP_035163.1 | 1 | 706 | ||
| GenBank mRNA | AK077154.1 | - | 1 | 706 | ||
| AK131715.1 | - | 1 | 685 | |||
| BC051134.1 | - | 1 | 686 | 6704229 | ||
| X75959.1 | - | 1 | 682 | |||

Pabpc2
Genomic Map